ContactUsLinkComponent
Qualitative filter papers, standard grades, grade 602 eh and 602 eh ½, Whatman™
Qualitative filter papers, standard grades, grade 602 eh and 602 eh ½, Whatman™
Certificates
undefined
Qualitative filter papers, standard grades, grade 602 eh and 602 eh ½, Whatman™
Specifications
  • Catalog No:
  • 515-5905
  • 515-5904
  • Grade:
  • 602 eh ½
  • 602 eh ½
  • Weight:
  • 85 g/m²
  • 85 g/m²
  • Thickness:
  • 150 µm
  • 150 µm
  • Particle retention:
  • <2 µm
  • <2 µm
  • Filtration speed:
  • 3000 s (Herzberg)
  • 3000 s (Herzberg)