Quinoline ≥98%
About this item
Purity: 98% CAS No.: 91-22-5 Molecular Formula: C9H7N Formula Weight: 129.16 Flash Point: 101°(214°F) Physical Form: Liquid Density: 1.090 Refractive Index: 1.6260 Sensitivity: Light Sensitive and Hygroscopic Appearance: Colourless to pale yellow pH: 7.3 Melting Point: -15° Boiling Point: 237° Beilstein Reference: 107477 Merck Reference: 14,8068 UN No.: 2656 MDL No.: MFCD00006736
- CAS Number:91-22-5
- Formula:C₉H₇N
- MDL Number:MFCD00006736
- Molecular Weight:129.16 g/mol
- Merck Index:14,8068
- Boiling Pt:237 °C
- Melting Pt:–15 °C
- Density:1.090
- Flash Pt:101 °C (214 °F)
- Storage Temperature:Ambient
- EINECS:202-051-6
- UN:2656
- ADR:6.1,III
3 Options Available Below
Product Details & Documents
Purity: 98% CAS No.: 91-22-5 Molecular Formula: C9H7N Formula Weight: 129.16 Flash Point: 101°(214°F) Physical Form: Liquid Density: 1.090 Refractive Index: 1.6260 Sensitivity: Light Sensitive and Hygroscopic Appearance: Colourless to pale yellow pH: 7.3 Melting Point: -15° Boiling Point: 237° Beilstein Reference: 107477 Merck Reference: 14,8068 UN No.: 2656 MDL No.: MFCD00006736
- CAS Number:91-22-5
- Formula:C₉H₇N
- MDL Number:MFCD00006736
- Molecular Weight:129.16 g/mol
- Merck Index:14,8068
- Boiling Pt:237 °C
- Melting Pt:–15 °C
- Density:1.090
- Flash Pt:101 °C (214 °F)
- Storage Temperature:Ambient
- EINECS:202-051-6
- UN:2656
- ADR:6.1,III



