Screw caps and pouring rings for laboratory bottles
Supplier: Kavalier
Certificates
About this item
Made of PP or PBT
- For DIN GL threads
- Heat resistant up to 140 °C (PP) and up to 180 °C (PBT)
- Autoclavable
Frequently Bought Together
Test strips, nitrate/nitrite, Dosatest
Supplier: VWR Chemicals
VWR®, Beakers, Tall Form
Supplier: VWR Collection
Potassium chloride, EMSURE® for analysis, Supelco®
Supplier: Merck
Chlorine, sensitive test strips, Dosatest
Supplier: VWR Chemicals
VWR®, Laboratory Bottles, Narrow Neck, with Screw Cap
Supplier: VWR Collection
VWR®, Powder packs for determination of chlorine, reagents for photometers
Supplier: VWR Chemicals
Screw caps and pouring rings for laboratory bottles
Supplier: Brand
2-Propanol, dehydrated (max. 0.01% H₂O) ≥99.8%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
VWR®, Aluminium foil in dispensers
Supplier: VWR Collection
DURAN® Screw Caps with Aperture
Supplier: DWK Life Sciences
Screw caps and pouring rings for laboratory bottles
Supplier: Kavalier
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Pouring rings for laboratory bottles, DURAN®
Supplier: DWK Life Sciences
Cyclohexanone ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Cell culture flasks, Corning®
Supplier: Corning
Spoon spatulas, for analysis
Supplier: BOCHEM
Screw caps for laboratory bottles, DURAN®
Supplier: DWK Life Sciences
Screw caps and pouring rings for SVS-components
Supplier: witeg Labortechnik
Screw caps with aperture
Supplier: Bohlender