Order Entry
Belgium
Orders LinkContactUsLinkComponent
Mesitylene 97%, pure

Mesitylene 97%, pure

Irritant
Environment
Flammable
Health
Mesitylene 97%, pure
Certificates
Mesitylene 97%, pure

3 Options Available Below

Product Details & Documents
Synonyms:  1,3,5-Trimethylbenzene
  • CAS Number:108-67-8
  • Formula:C₉H₁₂
  • MDL Number:MFCD00008538
  • Molecular Weight:120.19 g/mol
  • Merck Index:13,05934
  • Boiling Pt:163…166 °C (760 mmHg)
  • Melting Pt:–45 °C
  • Density:0.8675 g/cm³
  • Flash Pt:44 °C
  • Storage Temperature:Ambient
  • EINECS:203-604-4
  • UN:2325
  • ADR:3,III

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4980 to 1.5000 (20°C, 589 nm)
  • GC:
    ≥96.0 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 48,30 €
    48,30 €
    1 * 250 mL
       48,30 €

  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 119,00 €
    119,00 €
    1 * 1 L
       119,00 €

  • Glass bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L - 196,00 €
    196,00 €
    1 * 2,5 L
       196,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.