2,6-Di-tert-butyl-p-cresol for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Butylated hydroxytoluene, Butylhydroxytoluene, 2,6-Di-tert-butyl-4-methylphenol, BHT
CAS Number:128-37-0
:
Formula:C₁₅H₂₄O
MDL Number:MFCD00011644
:220.35 g/mol
Merck Index:13,01547
Boiling Pt:265 °C (1013 hPa)
Melting Pt:69…70 °C
Density:1.05 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:204-881-4
UN:3077
ADR:9,III
:M-Clarity™ quality level = MQ200
Flash Pt:127 °C
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:- 0.001 g/l (25 °C)
- Melting Point:69 - 70 °C
- Molar Mass:220.35 g/mol
- Bulk Density:450 kg/m3
- Boiling Point:265 °C (1013 hPa)
- Vapor Pressure:0.02 hPa (20 °C)
- Flash Point:127 °C
- Density:1.05 g/cm3 (20 °C)
- pH:(H2O) not applicable
- Ignition Point:345 °C
Specification Test Results
Related Information
Frequently Bought Together