2,6-Di-tert-butyl-p-cresol
EHERC12253500
:
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
:
:220.35 g/mol
:13,01547
:265 °C (1013 hPa)
:69…70 °C
:1.05 g/cm³ (20 °C)
:127 °C
:Ambient
:204-881-4
:128-37-0
:C₁₅H₂₄O
:MFCD00011644