Screw caps for screw thread tubes
Supplier: Lenz Laborglas
Jump to:
About this item
PBT,red, with PTFE coated silicone sealing (VMQ).
- For DIN GL threads
- Heat resistant up to 180 °C
- Autoclavable, sterilisable with hot air
5 Options Available Below
Product Details & Documents
PBT,red, with PTFE coated silicone sealing (VMQ).
- For DIN GL threads
- Heat resistant up to 180 °C
- Autoclavable, sterilisable with hot air
Product Family Options
Product Information