Order Entry
Belgium
Orders LinkContactUsLinkComponent
o-Xylene ≥99%, GPR RECTAPUR®

o-Xylene ≥99%, GPR RECTAPUR®

Flammable
Irritant
Supplier:   VWR Chemicals
o-Xylene ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
o-Xylene ≥99%, GPR RECTAPUR®

2 Options Available Below

Product Details & Documents
Synonyms:  1,2-Dimethylbenzene
  • CAS Number:95-47-6
  • Formula:C₈H₁₀
  • MDL Number:MFCD00008519
  • Molecular Weight:106.17 g/mol
  • Merck Index:12,10214
  • Boiling Pt:144 °C (1013 hPa)
  • Melting Pt:–25 °C
  • Density:0.88 g/cm³ (20 °C)
  • Flash Pt:30 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:202-422-2
  • UN:1307
  • ADR:3,III
Specifications for Product Family

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 99 %
  • IR Spectrum:
    Passes test
  • Density (20/4):
    0.870 - 0.890
Product Family Options

Product Information