o-Xylene ≥96% for spectrophotometry
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 1,2-Dimethylbenzene
CAS Number:95-47-6
:
Formula:C₈H₁₀
MDL Number:MFCD00008519
:106.17 g/mol
Merck Index:14,10081
Boiling Pt:143…145 °C
Melting Pt:–25 °C
Density:0.877
Flash Pt:32 °C (89 °F)
Storage Temperature:Ambient
EINECS:202-422-2
UN:1307
ADR:3,III
:GHS02
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together