Orders LinkContactUsLinkComponent
Microtubes
Microtubes
Supplier:   RATIOLAB
Microtubes
Supplier:  RATIOLAB
Certificates
Microtubes
4 Options Available Below
Product Details & Documents
Translucent polypropylene (PP) with very low protein binding.
  • Very good temperature resistance
  • Very good chemical resistance
The 0.65 and 1.2 ml micro-tubes can be supplied as single, strips of 8 or in 96-well format as a refill system for 8 x 12 plates.
Product Family Options
Product Information
  • Capacity
    Availability
    Price
  • RATI8200001
    single, for use with racks
    single, for use with racks
    1,2 ml
    Log in for availability
    Log in for availability
    1 * 5.000 ST
    1 * 5.000 ST - 274,00 €
    274,00 €
    1 * 5.000 ST
       274,00 €
  • RATI8200002
    strips of 8 tubes
    strips of 8 tubes
    1,2 ml
    Log in for availability
    Log in for availability
    1 * 600 ST
    1 * 600 ST - 283,00 €
    283,00 €
    1 * 600 ST
       283,00 €
  • RATI8200021
    single, for use with racks
    single, for use with racks
    0,65 ml
    Log in for availability
    Log in for availability
    1 * 5.000 ST
    1 * 5.000 ST - 274,00 €
    274,00 €
    1 * 5.000 ST
       274,00 €
  • RATI8200022
    strips of 8 tubes
    strips of 8 tubes
    0,65 ml
    Log in for availability
    Log in for availability
    1 * 600 ST
    1 * 600 ST - 283,00 €
    283,00 €
    1 * 600 ST
       283,00 €