ContactUsLinkComponent
Inoculating loops and needles
Inoculating loops and needles
Catalog # RATI7020200
Supplier:  RATIOLAB
undefined
Inoculating loops and needles
Catalog # RATI7020200
Supplier:  RATIOLAB
Specifications
  • Packed:
    100 peel packs of 20
  • Capacity:
    10 µl
  • Description:
    Loop
  • Pk:
    2.000
Specifications
About this item
Made of polystyrene (PS).
  • Hexagonal handle for safe handling
  • Beta-sterile
  • Sterile, individually packed in easy to open peel-packs