Orders LinkContactUsLinkComponent
PCR racks
PCR racks
Supplier:   RATIOLAB
PCR racks
Supplier:  RATIOLAB
Certificates
PCR racks
4 Options Available Below
Product Details & Documents
Racks made of PP and are characterised by their very thin but uniform walls.
  • 8×12 positions, micro test plate format
  • Autoclavable
  • Available in different colors for the convenient identification of different types of samples
  • Transparent lids available in two different heights (Lids must be ordered seperately)
Work and storage rack for 0,2 ml and 0,5 ml PCR tubes, PCR strips, and PCR segments.
Product Family Options
Product Information
  • Colour
    Availability
    Price
  • RATI6114100
    96-Well PCR rack
    96-Well PCR rack
    White
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 24,50 €
    24,50 €
    1 * 10 ST
       24,50 €
  • RATI6114102
    96-Well PCR rack
    96-Well PCR rack
    Blue
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 20,70 €
    20,70 €
    1 * 10 ST
       20,70 €
  • RATI6114103
    96-Well PCR rack
    96-Well PCR rack
    Green
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 20,70 €
    20,70 €
    1 * 10 ST
       20,70 €
  • RATI6114104
    96-Well PCR rack
    96-Well PCR rack
    Red
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 24,50 €
    24,50 €
    1 * 10 ST
       24,50 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Lid, transparent, height 14 mm
    Lid, transparent, height 14 mm
    RATI6114110
    Log in for availability
    1 * 10 ST - 25,10 €
Product Information
  • Catalog #
    Availability
    Price
  • Lid, transparent, height 28 mm
    Lid, transparent, height 28 mm
    RATI6114111
    Log in for availability
    1 * 10 ST - 28,20 €