Order Entry
Belgium
ContactUsLinkComponent
 
 
 
 
 
Isopentyl nitrite (mixed isomers ) 96%, Sigma-Aldrich®
 
undefined
 
 
 
 
Isopentyl nitrite (mixed isomers ) 96%, Sigma-Aldrich®

 

  • Appearance (Colour):
    Colorless to Yellow and Colourless to Green-Yellow
  • Appearance (Form):
    Liquid
  • Purity (GC AREA %):
    ≥95.5 %
  • Infrared spectrum:
    Conforms to structure

 

Frequently Bought Together