Order Entry
Belgium
ContactUsLinkComponent
 
Toxic
Health
Potassium dichromate 1/6 mol/l (1 N) in aqueous solution volumetric solution
Catalog # REGEKC2016F
Supplier:  REAGECON
CAS Number:  
Toxic
Health
Potassium dichromate 1/6 mol/l (1 N) in aqueous solution volumetric solution
Catalog # REGEKC2016F
Supplier:  REAGECON
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :294.18 g/mol
Storage Temperature:Ambient
EINECS:231-906-6
UN:3287
ADR:6.1,III
 :GHS06
 :GHS08
CAS Number:7778-50-9
Formula:K₂Cr₂O₇
MDL Number:MFCD00011367