Order Entry
Belgium
Orders LinkContactUsLinkComponent
Urea 40% in aqueous solution

Urea 40% in aqueous solution

10
Catalog # OXOISR0020K
Urea 40% in aqueous solution
Catalog # OXOISR0020K
CAS Number:  57-13-6
Urea 40% in aqueous solution
Urea 40% in aqueous solution

10

CAS Number:  57-13-6
Price
61,20 €
Log in for availability
1 * 10 ST
Product Details & Documents
Synonyms:  Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide
  • CAS Number:57-13-6
  • Formula:NH₂CONH₂
  • MDL Number:MFCD00008022
  • Molecular Weight:60.06 g/mol
  • Storage Temperature:Ambient
  • EINECS:200-315-5
Specifications

Specifications

  • Pk:
    10

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.