


Hydrogen peroxide-urea, Sigma-Aldrich®
: Merck
:124-43-6
:
:CH₆N₂O₃
:MFCD00013119
:94.07 g/mol
:5.1 (8),III
:M-Clarity™ quality level = MQ200
:204-701-4
:1511
:GHS03
:GHS05
:GHS07
:Ambient
- Solubility:500 g/l (20 °C)
- Melting Point:75 - 85 °C (decomposition)
- Molar Mass:94.07 g/mol
- pH:6.3 (H2O, 20 °C)
Frequently Bought Together