Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Galactitol 99+%

Galactitol 99+%
Certificates
Galactitol 99+%

3 Options Available Below

Product Details & Documents
Synonyms:  Dulcitol, rel-(2R,3S,4R,5S)-Hexane-1,2,3,4,5,6-hexaol
  • CAS Number:608-66-2
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00064288
  • Molecular Weight:182.17 g/mol
  • Boiling Pt:275…280 °C (1 mmHg)
  • Melting Pt:188…189 °C
  • Storage Temperature:Ambient
  • EINECS:210-165-2

 

Specifications for Product Family

Specification Test Results

  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    187 to 191 °C
  • Water:
    ≤1.0 % (K.F.)
  • Solubility:
    (3 % in water) Colorless
  • HPLC:
    ≥99.0 % (ELSD)
  • Heavy metals (as Pb):
    ≤0.003 %
  • Insoluble matter:
    ≤0.01 % (in dilute NH4OH)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 37,80 €
    37,80 €
    1 * 25 g
       37,80 €

  • Glass bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 93,50 €
    93,50 €
    1 * 100 g
       93,50 €

  • Glass bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 299,00 €
    299,00 €
    1 * 500 g
       299,00 €