Accessories for vortex mixers, TX4, ZX3, CLASSIC and WIZARD
Supplier: VELP SCIENTIFIC
Certificates
About this item
Frequently Bought Together
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Accessories for vortex mixers, TX4, ZX3, CLASSIC and WIZARD
Supplier: VELP SCIENTIFIC
VWR®, Laboratory Coats, Howie
Supplier: VWR Collection
VWR®, Digital Timer, One Channel
Supplier: VWR Collection
VWR®, Digital Timer, Four Channels, with Clock
Supplier: VWR Collection
Pipette tips, epT.I.P.S.® Standard (General Lab Product)
Supplier: EPPENDORF
Modular Basket System
Supplier: ELMA SCHMIDBAUER
VWR®, Nitrile Gloves
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Multipurpose containers
Supplier: Greiner Bio-One
Buffer solutions in sachets
Supplier: Mettler - Toledo
Kits for Precellys®
Supplier: Bertin Instruments