Orders LinkContactUsLinkComponent
Measuring cylinders tall form, without base, DURAN®
Supplier:   DWK Life Sciences
Measuring cylinders tall form, without base, DURAN®
Supplier:  DWK Life Sciences
Certificates
Measuring cylinders tall form, without base, DURAN®
Measuring cylinders tall form, without base, DURAN®
7 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
  • Tall form
  • Flat bottom
  • With spout
  • Without base
  • Without graduation
  • Very good chemical resistance
  • Very good temperature resistance
  • Without foot
Product Family Options
Product Information
  • Capacity
    Availability
    Price
  • 10 ml
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 243,00 €
    243,00 €
    1 * 50 ST
       243,00 €
  • 25 ml
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 70,60 €
    70,60 €
    1 * 10 ST
       70,60 €
  • 50 ml
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 45,30 €
    45,30 €
    1 * 10 ST
       45,30 €
  • 100 ml
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 119,00 €
    119,00 €
    1 * 20 ST
       119,00 €
  • 250 ml
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 149,00 €
    149,00 €
    1 * 10 ST
       149,00 €
  • 500 ml
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 264,00 €
    264,00 €
    1 * 10 ST
       264,00 €
  • 1000 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 85,20 €
    85,20 €
    1 * 1 ST
       85,20 €