Orders LinkContactUsLinkComponent
Sedimentation cones, Imhoff type, DURAN®
Sedimentation cones, Imhoff type, DURAN®
Imhoff cones 1000 ml, without stopcock
Catalog # SCOT214005402
Supplier:   DWK Life Sciences
Sedimentation cones, Imhoff type, DURAN®
Catalog # SCOT214005402
Supplier:  DWK Life Sciences
Sedimentation cones, Imhoff type, DURAN®
Sedimentation cones, Imhoff type, DURAN®
Imhoff cones 1000 ml, without stopcock
Price
1 130,00 €
Log in for availability
1 * 10 ST
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
  • Without graduations
  • Very good chemical resistance
  • Very good temperature resistance
Specifications
  • Type:
    without stopcock
  • Description:
    Imhoff cones 1000 ml