Order Entry
Belgium
ContactUsLinkComponent
Toluene ≥99.7%, PESTINORM® for pesticide residue analysis
Toluene ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier:  VWR Chemicals
Certificates
undefined
Toluene ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 99.7 %
  • Evaporation residue:
    Max. 5 ppm
  • Water:
    Max. 0.03 %
  • Lindane (GC/ECD) ≤ 5 ng/l:
    Passes test
  • Pesticide analysis (Lindane/ECD):
    Max. 5 ng/l

Specification Test Results

Frequently Bought Together