Order Entry
Belgium
ContactUsLinkComponent
Toluene ≥98%, TECHNICAL
Flammable
Health
Irritant
Toluene ≥98%, TECHNICAL
Supplier:  VWR Chemicals
Certificates
undefined
Flammable
Health
Irritant
Toluene ≥98%, TECHNICAL
Supplier:  VWR Chemicals
Certificates
Limited Time Sale
Select items below are currently on sale

Specification Test Results

  • Assay:
    Min. 98 %

Specification Test Results

Frequently Bought Together