Order Entry
Belgium
ContactUsLinkComponent
Toluene, EMPARTA® ACS analytical reagent, Supelco®
Flammable
Health
Irritant
Toluene, EMPARTA® ACS analytical reagent, Supelco®
Supplier:  Merck
Certificates
undefined
Flammable
Health
Irritant
Toluene, EMPARTA® ACS analytical reagent, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    0.52 g/l (20 °C)
  • Melting Point:
    -95 °C
  • Molar Mass:
    92.14 g/mol
  • Boiling Point:
    110.6 °C (1013 hPa)
  • Vapor Pressure:
    29 hPa (20 °C)
  • Flash Point:
    4 °C
  • Refractive Index:
    1.4968 (20 °C)
  • Explosion Limit:
    1.2 - 8 %(V)
  • Density:
    0.87 g/cm3 (20 °C)
  • pH:
    (H2O) not applicable
  • Ignition Point:
    535 °C
  • Viscosity Kinematic:
    0.7 mm2/s (20 °C)

Specification Test Results

Related Information

Frequently Bought Together