Order Entry
Belgium
ContactUsLinkComponent
Toluene, GPR RECTAPUR® low in sulphur
Flammable
Health
Irritant
Toluene, GPR RECTAPUR® low in sulphur
Catalog # 304547S
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Health
Irritant
Toluene, GPR RECTAPUR® low in sulphur
Catalog # 304547S
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

About this item

 :
 :92.14 g/mol
Merck Index:13,09607
Boiling Pt:110.6 °C (1013 hPa)
Melting Pt:–95 °C
Density:0.867 g/cm³ (20 °C)
Flash Pt:4 °C
Storage Temperature:Ambient
EINECS:203-625-9
UN:1294
ADR:3,II
REACH:05-2114615130-69
 :GHS02
 :GHS08
 :GHS07
CAS Number:108-88-3
Formula:C₆H₅CH₃
MDL Number:MFCD00008512