Order Entry
Belgium
ContactUsLinkComponent
Toluene, low water content ≥99.5% (by GC, corrected for water content), BakerDRY™, J.T.Baker®
Toluene, low water content ≥99.5% (by GC, corrected for water content), BakerDRY™, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Toluene, low water content ≥99.5% (by GC, corrected for water content), BakerDRY™, J.T.Baker®
Supplier:  Avantor

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    ≥ 99.5 %
  • ACS - Assay (C₆H₅CH₃) (by GC, corrected for water):
    ≥99.5 %
  • ACS - Color (APHA):
    ≤10
  • ACS - Residue after Evaporation:
    ≤1 ppm
  • ACS - Substances Darkened by H₂SO₄:
    Passes Test
  • ACS - Sulfur Compounds (as S):
    ≤0.003 %
  • ACS - Water (by KF, coulometric):
    ≤10.0 ppm

Specification Test Results

Frequently Bought Together