Orion™ RDO™ Dissolved Oxygen Probes
Supplier: Thermo Orion
About this item
These RDO (Rugged Dissolved Oxygen) probes use optical DO technology for problem-free measurement.
- Measure dissolved oxygen levels without the difficulties of membrane replacement
- Once-a-year maintenance with a simple cap replacement
- Different cable lengths available
- IP 68-rated, waterproof
Can be used with Orion Star meters with RDO input.
: Supplied with optical cap, stainless steel sensor guard and calibration sleeve.
2 Options Available Below
Product Details & Documents
These RDO (Rugged Dissolved Oxygen) probes use optical DO technology for problem-free measurement.
- Measure dissolved oxygen levels without the difficulties of membrane replacement
- Once-a-year maintenance with a simple cap replacement
- Different cable lengths available
- IP 68-rated, waterproof
Can be used with Orion Star meters with RDO input.
: Supplied with optical cap, stainless steel sensor guard and calibration sleeve.
Specifications for Product Family
Specifications
- Catalog No:
- 664-0154
- 664-0155
- Probe type:
- RDO optical dissolved oxygen
- RDO optical dissolved oxygen
- Sensor type:
- RDO optical dissolved oxygen sensor
- RDO optical dissolved oxygen sensor
Specifications
Product Family Options
Product Information
- TypeConcentrationTemp.ConnectionØ×LForAvailabilityPrice
- 664-0154Orion™ RDO™ optical DO probeOrion™ RDO™ optical DO probeRDO™ 087010MD0 - 20 mg/L, 0 - 200%0...50 °C3 m cable16/33×70/190 mmOrion Star Meter with RDO inputLog in for availability1 * 1 ST1 240,00 €Specifications:
More
Specifications
Cat. No.664-0154Probe typeRDO optical dissolved oxygenSensor typeRDO optical dissolved oxygen sensor - 664-0155Orion™ RDO™ optical DO probeOrion™ RDO™ optical DO probeRDO™ 087020MD0 - 20 mg/L, 0 - 200%0...50 °C6 m cable16/33×70/190 mmOrion Star Meter with RDO inputLog in for availability1 * 1 ST1 290,00 €Specifications:
More
Specifications
Cat. No.664-0155Probe typeRDO optical dissolved oxygenSensor typeRDO optical dissolved oxygen sensor