Cell counter vials
Supplier: RATIOLAB
Aller à :
À propos de cet article
Cell counter vials made of clear PS.
- High-precision design for use with robotic analytical systems
4 Options disponibles ci-dessous
Product Details & Documents
Cell counter vials made of clear PS.
- High-precision design for use with robotic analytical systems
Product Family Options
Informations sur le produit
- CapacityMaterialColourCap includedPackedAvailabilityPrice
- RATI5515010Cell counter vial 1 with lid, for Coulter® and Hycel® cell counter systemsCell counter vial 1 with lid, for Coulter® and Hycel® cell counter systems25,00 mlPSClearYes50 vials/tray, 20 traysConnexion Pour la disponibilité1 * 1.000 ST127,00 €
- RATI5515040Cell counter vial 1 with lid, for Coulter® and Hycel® cell counter systemsCell counter vial 1 with lid, for Coulter® and Hycel® cell counter systems25,00 mlPSClearYes250 vials/bag, 4 bagsConnexion Pour la disponibilité1 * 1.000 ST109,00 €
- RATI5515041Cell counter vial 1 without lid, for Coulter® and Hycel® cell counter systemsCell counter vial 1 without lid, for Coulter® and Hycel® cell counter systems25,00 mlPSClearNo250 vials/bag, 4 bagsConnexion Pour la disponibilité1 * 1.000 ST68,70 €
- RATI5515051Cell counter vial 2 without lid, for cell counter systems with cylindrical containers, e.g. Al® and Linson®Cell counter vial 2 without lid, for cell counter systems with cylindrical containers, e.g. Al® and Linson®20,00 mlPSClearNo1000 vials/bagConnexion Pour la disponibilité1 * 1.000 ST137,00 €
Closures
Informations sur le produit
- Catalog #DescriptionAvailabilityPrice