Order Entry
Belgium
Orders LinkContactUsLinkComponent
Scintillation vials

Scintillation vials

20,00 ml, 27×60 mm, Yes
Catalog # 218-2255
Supplier:   RATIOLAB
Scintillation vials
Catalog # 218-2255
Supplier:  RATIOLAB
Scintillation vials
Scintillation vials

20,00 ml, 27×60 mm, Yes

Price
160,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

Scintillation vials made of polyethylene (PE-LD).

  • Convenient closing and opening
  • Leakproof cap
  • For use with robotic analytical system
Specifications
  • Cap included:
    Yes
  • Ø×H:
    27×60 mm
  • Capacity:
    20,00 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.