Toluene, extra dry over Molecular Sieve 99.85%, AcroSeal®
:108-88-3
:
:C₆H₅CH₃
:MFCD00008512
:92.14 g/mol
:13,09607
:111 °C (760 mmHg)
:–95 °C
:0.866 g/cm³
:4 °C
:Ambient
:203-625-9
:1294
:3,II
:05-2114615130-69
:GHS07
:GHS08
:GHS02
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Water:≤0.005 % (Coulometric)
- GC:≥99.8 %
- Acidity (HCl):≤0.002 %
Frequently Bought Together