Orders LinkContactUsLinkComponent
Stopcocks, single way, DURAN®
Stopcocks, single way, DURAN®
Supplier:   DWK Life Sciences
Stopcocks, single way, DURAN®
Supplier:  DWK Life Sciences
Certificates
Stopcocks, single way, DURAN®
Stopcocks, single way, DURAN®
2 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear, with hollow glass key and glass handle.
  • Replaceable NS keys
  • Screw thread retaining device
Straight bore.
Product Family Options
Product Information
  • Hole Ø
    Spigot
    Availability
    Price
  • 2,5 mm
    14,5 NS
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 1 940,00 €
    1 940,00 €
    1 * 50 ST
       1 940,00 €
  • 4 mm
    18,8 NS
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 1 120,00 €
    1 120,00 €
    1 * 25 ST
       1 120,00 €