Stopcocks, single-way, DURAN®
Supplier: DWK Life Sciences
Jump to:
About this item
DURAN®, borosilicate glass 3.3, clear, with glass or PTFE keys.
- Replaceable NS keys
- Screw thread retaining device
Straight bore.
19 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear, with glass or PTFE keys.
- Replaceable NS keys
- Screw thread retaining device
Straight bore.
Product Family Options
With SBW glass key
Product Information
With grooved SBW glass key
Product Information
- Hole ØSpigotAvailabilityPrice
With PTFE key
Product Information
- Hole ØSpigotAvailabilityPrice
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.