Order Entry
Belgium
Orders LinkContactUsLinkComponent
Storage Box for Sample Tubes

Storage Box for Sample Tubes

Supplier:   Simport Scientific
Storage Box for Sample Tubes
Supplier:  Simport Scientific
Certificates
Storage Box for Sample Tubes
Storage Box for Sample Tubes

16 Options Available Below

Product Details & Documents

Cover and base made of polystyrene.

  • Sturdy
  • Stackable
  • Wide variety of economical storage boxes

Printed with a series of numbered squares and the surface accepts the writing with markers by facilitating inventory control.

These storage boxes are designed with a transparent cover that allows the user to see the contents of the box and is keyed to the base to prevent misalignment.

Product Family Options

25 well racks

Product Information

  • No. of Holes
    Colour
    Dimensions
    For tubes
    Availability
    Price
  • 25 (5×5)
    Blue
    76×76×52 mm
    1 - 2 ml
    Log in for availability
    Log in for availability
    1 * 48 ST
    1 * 48 ST - 298,00 €
    298,00 €
    1 * 48 ST
       298,00 €

  • 25 (5×5)
    Green
    76×76×52 mm
    1 - 2 ml
    Log in for availability
    Log in for availability
    1 * 48 ST
    1 * 48 ST - 298,00 €
    298,00 €
    1 * 48 ST
       298,00 €

  • 25 (5×5)
    Pink
    76×76×52 mm
    1 - 2 ml
    Log in for availability
    Log in for availability
    1 * 48 ST
    1 * 48 ST - 298,00 €
    298,00 €
    1 * 48 ST
       298,00 €

  • 25 (5×5)
    Yellow
    76×76×52 mm
    1 - 2 ml
    Log in for availability
    Log in for availability
    1 * 48 ST
    1 * 48 ST - 298,00 €
    298,00 €
    1 * 48 ST
       298,00 €

81 well racks

Product Information

  • No. of Holes
    Colour
    Dimensions
    For tubes
    Availability
    Price
  • 81 (9×9)
    Blue
    133×133×52 mm
    1 - 2 ml
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 223,00 €
    223,00 €
    1 * 24 ST
       223,00 €

  • 81 (9×9)
    Green
    133×133×52 mm
    1 - 2 ml
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 223,00 €
    223,00 €
    1 * 24 ST
       223,00 €

  • 81 (9×9)
    Pink
    133×133×52 mm
    1 - 2 ml
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 223,00 €
    223,00 €
    1 * 24 ST
       223,00 €

  • 81 (9×9)
    Yellow
    133×133×52 mm
    1 - 2 ml
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 223,00 €
    223,00 €
    1 * 24 ST
       223,00 €

  • 81 (9×9)
    Blue
    133×133×95 mm
    3 - 5 ml
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 114,00 €
    114,00 €
    1 * 10 ST
       114,00 €

  • 81 (9×9)
    Green
    133×133×95 mm
    3 - 5 ml
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 114,00 €
    114,00 €
    1 * 10 ST
       114,00 €

  • 81 (9×9)
    Pink
    133×133×95 mm
    3 - 5 ml
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 114,00 €
    114,00 €
    1 * 10 ST
       114,00 €

  • 81 (9×9)
    Yellow
    133×133×95 mm
    3 - 5 ml
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 111,00 €
    111,00 €
    1 * 10 ST
       111,00 €

100 well racks

Product Information

  • No. of Holes
    Colour
    Dimensions
    For tubes
    Availability
    Price
  • 100 (10×10)
    Blue
    133×133×52 mm
    1 - 2 ml
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 213,00 €
    213,00 €
    1 * 24 ST
       213,00 €

  • 100 (10×10)
    Green
    133×133×52 mm
    1 - 2 ml
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 213,00 €
    213,00 €
    1 * 24 ST
       213,00 €

  • 100 (10×10)
    Pink
    133×133×52 mm
    1 - 2 ml
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 213,00 €
    213,00 €
    1 * 24 ST
       213,00 €

  • 100 (10×10)
    Yellow
    133×133×52 mm
    1 - 2 ml
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 213,00 €
    213,00 €
    1 * 24 ST
       213,00 €