Multi purpose bottles
Supplier: RATIOLAB
Aller à :
À propos de cet article
Polyethylene, blue translucent.
- Incl. lid with nonwoven fabric liner
- With graduations
- Light protection
- Wide neck diameter of 80 mm
3 Options disponibles ci-dessous
Product Details & Documents
Polyethylene, blue translucent.
- Incl. lid with nonwoven fabric liner
- With graduations
- Light protection
- Wide neck diameter of 80 mm
Product Family Options
Informations sur le produit
- CapacityColourNeck-ØAvailabilityPrice
- RATI5922250Multipurpose collecting bottles with labelMultipurpose collecting bottles with label2500 mlBlue80 mmConnexion Pour la disponibilité1 * 60 ST205,00 €
- RATI5922204Lids for multipurpose collecting bottles, 2000 or 2500 ml capacityLids for multipurpose collecting bottles, 2000 or 2500 ml capacityConnexion Pour la disponibilité1 * 100 ST
- RATI5922253Labels, self-adhesive, for multi purpose bottles