Orders LinkContactUsLinkComponent
Stoppers, for test tubes
Stoppers, for test tubes
Supplier:   RATIOLAB
Stoppers, for test tubes
Supplier:  RATIOLAB
Certificates
Stoppers, for test tubes
Stoppers, for test tubes
5 Options Available Below
Product Details & Documents
PE.
  • Very tight seal due to two sealing rings
  • Available in natural and four more colors
Product Family Options
Product Information
  • Colour
    Ø bottom
    Availability
    Price
  • Natural
    11 - 12 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 22,10 €
    22,10 €
    1 * 1.000 ST
       22,10 €
  • Blue
    11 - 12 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 22,10 €
    22,10 €
    1 * 1.000 ST
       22,10 €
  • Green
    11 - 12 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 22,10 €
    22,10 €
    1 * 1.000 ST
       22,10 €
  • Red
    11 - 12 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 22,10 €
    22,10 €
    1 * 1.000 ST
       22,10 €
  • Natural
    16 - 17 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 22,10 €
    22,10 €
    1 * 1.000 ST
       22,10 €