Orders LinkContactUsLinkComponent
Sample tubes
Sample tubes
Supplier:   Simport Scientific
Sample tubes
Supplier:  Simport Scientific
Certificates
Sample tubes
5 Options Available Below
Product Details & Documents
PP, translucent
  • Withstand temperatures from −196 to 121 °C
  • Withstand centrifugation
  • Autoclavable
  • Non sterile
For storing and transporting biological material. Tube with internal threads. Caps are supplied with or without a silicone washer, to ensure a positive leakproof seal at all temperatures. A 1 1/4 turn of the cap is sufficient to seal the vial.
Since both closures and tubes are manufactured of the same material, they have the same coefficient of expansion to guarantee an equally secure seal both at room or at low temperatures. Round bottom tubes only can be centrifuged up to 14000xg.
Ordering information: Order caps separately.
Product Family Options
Tubes without screw caps
Product Information
  • Capacity
    Size
    Availability
    Price
  • 2 ml
    12,5×49 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 124,00 €
    124,00 €
    1 * 1.000 ST
       124,00 €
  • 4 ml
    12,5×72 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 153,00 €
    153,00 €
    1 * 1.000 ST
       153,00 €
  • 4 ml
    12,5×72 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 166,00 €
    166,00 €
    1 * 1.000 ST
       166,00 €
  • 5 ml
    12,5×92 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 195,00 €
    195,00 €
    1 * 1.000 ST
       195,00 €
  • 1,2 ml
    12,5×43 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 107,00 €
    107,00 €
    1 * 1.000 ST
       107,00 €
Screw caps
Product Information
  • Catalog #
    Description
    Colour
    Availability
    Price
  • Screw caps with silicone washer
    Red
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 190,00 €
    190,00 €
    1 * 1.000 ST
       190,00 €
Product Information
  • Catalog #
    Description
    Colour
    Availability
    Price
  • Screw caps with silicone washer
    White
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 190,00 €
    190,00 €
    1 * 1.000 ST
       190,00 €
Product Information
  • Catalog #
    Description
    Colour
    Availability
    Price
  • Screw caps with silicone washer
    Yellow
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 190,00 €
    190,00 €
    1 * 1.000 ST
       190,00 €
Product Information
  • Catalog #
    Description
    Colour
    Availability
    Price
  • Screw caps with silicone washer
    Blue
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 190,00 €
    190,00 €
    1 * 1.000 ST
       190,00 €
Product Information
  • Catalog #
    Description
    Colour
    Availability
    Price
  • Screw caps with silicone washer
    Green
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 190,00 €
    190,00 €
    1 * 1.000 ST
       190,00 €
Product Information
  • Catalog #
    Description
    Colour
    Availability
    Price
  • Screw caps with silicone washer
    Natural
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 190,00 €
    190,00 €
    1 * 1.000 ST
       190,00 €