Orders LinkContactUsLinkComponent
Cryogenic tubes
Supplier:   RATIOLAB
Cryogenic tubes
Supplier:  RATIOLAB
Certificates
9 Options Available Below
Product Details & Documents
PP tubes with integrated seal in lid and white writing area. Developed for storing and conserving the tissue culture and biological samples at extremely low temperatures. Suitable for use in freezers as well as in gaseous phase of liquid nitrogen to -190 °C.
  • White graduated
  • Equipped with base for using with one hand
  • Free of DNA, DNase, pyrogens and ATP
Produced in a class 7 cleanroom (class 10000) according to DIN ISO 13485:2004, ISO 14644 and 14698 (Federal Standard 209).
 : Produced in a class 7 cleanroom (class 10000) according to DIN ISO 13485:2004, ISO 14644 and 14698 (Federal Standard 209).
 : Packaged in PE bags containing 100 tubes in boxes.
Product Family Options
With male thread
Product Information
  • Capacity
    Base
    Ø×H
    Cap included
    Availability
    Price
  • 1,20 ml
    Freestanding
    12,3×41 mm
    Yes
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 297,00 €
    297,00 €
    1 * 1.000 ST
       297,00 €
  • 2,00 ml
    Round bottom
    12,3×46 mm
    Yes
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 297,00 €
    297,00 €
    1 * 1.000 ST
       297,00 €
  • 2,00 ml
    Freestanding
    12,3×47 mm
    Yes
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 297,00 €
    297,00 €
    1 * 1.000 ST
       297,00 €
  • 3,00 ml
    Freestanding
    12,3×71 mm
    Yes
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 330,00 €
    330,00 €
    1 * 1.000 ST
       330,00 €
  • 4,00 ml
    Freestanding
    12,3×75 mm
    Yes
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 364,00 €
    364,00 €
    1 * 1.000 ST
       364,00 €
  • 5,00 ml
    Freestanding
    12,3×90 mm
    Yes
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 414,00 €
    414,00 €
    1 * 1.000 ST
       414,00 €
With female thread
Product Information
  • Capacity
    Base
    Ø×H
    Cap included
    Availability
    Price
  • 2,00 ml
    Freestanding
    12,3×48 mm
    Yes
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 330,00 €
    330,00 €
    1 * 1.000 ST
       330,00 €
  • 4,00 ml
    Freestanding
    12,3×76 mm
    Yes
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 347,00 €
    347,00 €
    1 * 1.000 ST
       347,00 €
  • 5,00 ml
    Freestanding
    12,3×91 mm
    Yes
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 446,00 €
    446,00 €
    1 * 1.000 ST
       446,00 €