4-Nitrotoluene
1
Catalog # PROOERM-FC029
Supplier: LGC Standards PROMOCHEM
Jump to:
Product Details & Documents
- CAS Number:99-99-0
- Formula:C₇H₇NO₂
- MDL Number:MFCD00007366
- Molecular Weight:137.14 g/mol
- Merck Index:12,06748
- Boiling Pt:238.3 °C (1013 hPa)
- Melting Pt:51 °C
- Density:1.39 g/cm³ (20 °C)
- Flash Pt:106 °C
- Storage Temperature:Ambient
- EINECS:202-808-0
- UN:3446
- ADR:6.1,II
Specifications
Specifications
- Pk:1
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



