Stopcocks, straight or multi-way bore
Supplier: BURKLE
Jump to:
About this item
PTFE.
- Resistant to compression max. 1 bar
- Vacuum suitable
- Autoclavable
6 Options Available Below
Product Details & Documents
PTFE.
- Resistant to compression max. 1 bar
- Vacuum suitable
- Autoclavable
Documents
BURK8610-0006_TDS_EN
Datasheet
Product Family Options
2-way Valve
Product Information
3-way Valve
Product Information