Orders LinkContactUsLinkComponent
Test tubes
Test tubes
Supplier:   RATIOLAB
Test tubes
Supplier:  RATIOLAB
Certificates
Test tubes
5 Options Available Below
Product Details & Documents
PS, clear or PP, translucent.
  • PS: Good thermal resistance. PP: Very good thermal resistance, autoclavable
  • PS: Good chemical resistance. PP: Very good chemical resistance
  • Robust, economical
Universal tubes for a wide range of applications.
Product Family Options
Round bottom
Product Information
  • Capacity
    Length
    Ext.Ø
    Material
    Packed
    Availability
    Price
  • 5 ml
    75 mm
    12 mm
    PS
    1000/bag
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 30,10 €
    30,10 €
    1 * 1.000 ST
       30,10 €
  • 4 ml
    70 mm
    11 mm
    PS
    1000/bag
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 33,80 €
    33,80 €
    1 * 1.000 ST
       33,80 €
  • 12 ml
    100 mm
    16 mm
    PS
    1000/bag
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 61,40 €
    61,40 €
    1 * 1.000 ST
       61,40 €
  • 5 ml
    75 mm
    12 mm
    PP
    1000/bag
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 31,90 €
    31,90 €
    1 * 1.000 ST
       31,90 €
  • 4 ml
    55 mm
    12 mm
    PS
    1000/bag
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 30,60 €
    30,60 €
    1 * 1.000 ST
       30,60 €