Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Sedimentation cones, Imhoff type

Imhoff cone, 1000 ml, with stopper
  BURK7118-1000
 :   BURKLE
Sedimentation cones, Imhoff type
  BURK7118-1000
 :  BURKLE
Sedimentation cones, Imhoff type
Sedimentation cones, Imhoff type

Imhoff cone, 1000 ml, with stopper

 
69,40 €
   
1 * 1 ST
 

Styrene-acrylonitrile (SAN), PP screw plug.

  • As transparent as glass
  • Easy draining via screw plug

Graduation: 0 to 2 ml:0,1/ 2 to 10 ml:0,5/ 10 to 40 ml:1/ 40 to 100 ml:2/ 100 to 1000 ml:10

 
  • Type:
    with stopper
  • Description:
    Imhoff cone, 1000 ml
Accessories

 

  •  
    Description
     
     
  • BURK7118-2000
    for two Imhoff funnels
       
    1 * 1 ST - 143,00 €