Orders LinkContactUsLinkComponent
Stoppers, flange plug caps
Stoppers, flange plug caps
Supplier:   Simport Scientific
Stoppers, flange plug caps
Supplier:  Simport Scientific
Certificates
Stoppers, flange plug caps
Stoppers, flange plug caps
11 Options Available Below
Product Details & Documents
PE Plug type push-in caps and 2-position snap caps are made of polyethylene for test tubes with outside diameters of 12 or 17 mm. Dual position caps offer two possibilities: the closed but unsealed position where samples are maintained aerobic for microbiological procedures; and the fully sealed position where the cap is pushed tight to seal the tube for anaerobic use or for storage, transfer and centrifuge applications.
Product Family Options
Product Information
  • Colour
    Ø bottom
    Availability
    Price
  • Blue
    12 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 53,80 €
    53,80 €
    1 * 1.000 ST
       53,80 €
  • Natural
    12 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 53,80 €
    53,80 €
    1 * 1.000 ST
       53,80 €
  • Green
    12 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 53,80 €
    53,80 €
    1 * 1.000 ST
       53,80 €
  • Red
    12 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 53,80 €
    53,80 €
    1 * 1.000 ST
       53,80 €
  • Yellow
    12 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 53,80 €
    53,80 €
    1 * 1.000 ST
       53,80 €
  • Natural
    16 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 73,50 €
    73,50 €
    1 * 1.000 ST
       73,50 €
  • Blue
    16 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 73,50 €
    73,50 €
    1 * 1.000 ST
       73,50 €
  • Green
    16 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 73,50 €
    73,50 €
    1 * 1.000 ST
       73,50 €
  • Red
    16 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 73,50 €
    73,50 €
    1 * 1.000 ST
       73,50 €
  • White
    16 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 73,50 €
    73,50 €
    1 * 1.000 ST
       73,50 €
  • Yellow
    16 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 73,50 €
    73,50 €
    1 * 1.000 ST
       73,50 €