ContactUsLinkComponent
Measuring beakers with handle
Measuring beakers with handle
Catalog # BURK7104-5000
Supplier:  BURKLE
Supplier #: 7104-5000
undefined
Measuring beakers with handle
Catalog # BURK7104-5000
Supplier:  BURKLE
Supplier #: 7104-5000
Log in for availability
Select an Option
210 mm
Select an Option
5000 ml
Select an Option
270 mm
Select an Option
11
Select an Option
1
About this item
PP, highly transparent.
  • Ergonomic handle for secure grip
  • Functional spout to minimize spills
  • Cleaning up to max. +60 °C is recommended to preserve marks and inscriptions, for autoclaving (+121 °C) beakers with an transparent scale should be selected
Ideal for general laboratory use and a wide variety of other applications.
Frequently Bought Together