Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Closures
Test/Sample Tube Closures
Caps for microcentrifuge tubes, tamper evident, Micrewtube®
Caps for microcentrifuge tubes, tamper evident, Micrewtube®
Caps for microcentrifuge tubes, tamper evident, Micrewtube®
SIMPT340NOSFTTP
:
Simport Scientific
Caps for microcentrifuge tubes, tamper evident, Micrewtube®
SIMPT340NOSFTTP
:
Simport Scientific
Colour:
Natural
Description:
Without loop
Pk:
1.000