Orders LinkContactUsLinkComponent
Disposable cuvettes
Disposable cuvettes
Supplier:  RATIOLAB
Certificates
Disposable cuvettes
2 Options Available Below
Product Details & Documents
These cuvettes with reduced wall thickness result in increased heat transfer.
  • Very low variation of extinction values
 : In PS boxes with lid, 100 cuvettes per box.
Product Family Options
Product Information
  • Type
    Range
    Height
    Volume
    Path length
    Window material
    Availability
    Price
  • Macro
    340 – 900 nm
    45 mm
    4 ml
    10 mm
    PS
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 114,00 €
    114,00 €
    1 * 1.000 ST
       114,00 €
  • Semi-micro
    340 – 900 nm
    45 mm
    1,6 ml
    10 mm
    PS
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 116,00 €
    116,00 €
    1 * 1.000 ST
       116,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Stoppers for cuvettes, PP
    Stoppers for cuvettes, PP
    RATI2812011
    Log in for availability
    1 * 1.000 ST - 54,80 €