Adapters, conical ground joint on DIN thread, DURAN®
Fournisseur: DWK Life Sciences
Certificats
À propos de cet article
DURAN®, borosilicate glass 3.3, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Threaded adapter with DIN thread and standard ground cone.
DIN 12257
* non-DIN size
Frequently Bought Together