

Potassium dichromate 0.15 mol/l (0.9 N) in aqueous solution
Catalog # VWRS6075.9010
Supplier: VWR Collection
CAS Number:
Specifications
- Pk:10 L
- Seq:5214
Specifications
About this item
:
:294.18 g/mol
Storage Temperature:Ambient
EINECS:231-906-6
UN:3287
ADR:6.1,III
:GHS06
:GHS08
CAS Number:7778-50-9
Formula:K₂Cr₂O₇
MDL Number:MFCD00011367