m-Toluic acid for synthesis
Supplier: Merck
Certificates
About this item
CAS Number:99-04-7
:
Formula:C₈H₈O₂
MDL Number:MFCD00002523
:136.15 g/mol
Merck Index:12,09673
Boiling Pt:263 °C (1013 hPa)
Melting Pt:108…111 °C
Density:1.054 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:150 °C
EINECS:202-723-9
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:1 g/l (20 °C)
- Melting Point:108 - 111 °C
- Molar Mass:136.15 g/mol
- Bulk Density:500 kg/m3
- Boiling Point:263 °C (1013 hPa)
- Flash Point:150 °C
- pH:3.4 (H2O, 20 °C) (saturated solution)
- Ignition Point:500 °C
Specification Test Results
Frequently Bought Together