m-Toluic acid 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:99-04-7
:
Formula:C₈H₈O₂
MDL Number:MFCD00002523
:136.15 g/mol
Merck Index:12,09673
Boiling Pt:263 °C (1013 hPa)
Melting Pt:108…112 °C
Density:1.054 g/cm³
Flash Pt:150 °C
EINECS:202-723-9
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:Light yellow to light beige Crystalline solid
- Infrared spectrum:Conforms
- Melting point:108 to 112 °C
- Titration with NaOH:≥98.5 %
Specification Test Results
Frequently Bought Together