Orders LinkContactUsLinkComponent
Stand bases
Stand bases
Supplier:   BOCHEM
Stand bases
Supplier:  BOCHEM
Certificates
Stand bases
4 Options Available Below
Product Details & Documents
Stand bases made of stainless steel 18/10 with expanded, flat and heavy feet for stability.
  • With M 10 threaded hole (off-centre)
  • One adjustable foot
  • Anti-slip rubber protection
Certifications: DIN 12892
Product Family Options
Product Information
  • Weight
    W×D×H
    Availability
    Price
  • 1,6 kg
    130×210×3 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 137,00 €
    137,00 €
    1 * 1 ST
       137,00 €
  • 1,9 kg
    160×250×3 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 153,00 €
    153,00 €
    1 * 1 ST
       153,00 €
  • 2,0 kg
    150×300×3 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 161,00 €
    161,00 €
    1 * 1 ST
       161,00 €
  • 2,4 kg
    200×315×3 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 242,00 €
    242,00 €
    1 * 1 ST
       242,00 €