Orders LinkContactUsLinkComponent
Flow meters
Flow meters
Supplier:   BURKLE
Flow meters
Supplier:  BURKLE
Certificates
Flow meters
3 Options Available Below
Product Details & Documents
These flow meters are compact and fully electronic for electric drum and container pumps, also used for low viscosity media.
  • Measurement by the turbine wheel principle
  • Can be calibrated depending on the medium
  • Different versions for many different applications
  • Automatic switch on when flow is recognized
  • Four resettable part ranges and one fixed total range
Product Family Options
Product Information
  • Flow rate
    Accuracy
    Material
    Pressure
    Availability
    Price
  • BURK5700-0111
    Flow meter for fuel oil, diesel, hydrochloric acid, sodium hydroxide with connector thread G 3/4"
    Flow meter for fuel oil, diesel, hydrochloric acid, sodium hydroxide with connector thread G 3/4"
    5 - 90 l/min
    ±1 %
    PP, Hastelloy C4, FKM
    max.4 bar
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 340,00 €
    1 340,00 €
    1 * 1 ST
       1 340,00 €
  • BURK5700-0121
    Flow meter for fuel oil, diesel, hydrochloric acid, sodium hydroxide with connector thread G 1 1/4"
    Flow meter for fuel oil, diesel, hydrochloric acid, sodium hydroxide with connector thread G 1 1/4"
    5 - 90 l/min
    ±1 %
    PP, Hastelloy C4, FKM
    max.4 bar
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 100,00 €
    1 100,00 €
    1 * 1 ST
       1 100,00 €
  • BURK5700-0141
    Flow meter for highly aggressive liquids with connector thread G 1 1/4"
    Flow meter for highly aggressive liquids with connector thread G 1 1/4"
    5 - 90 l/min
    ±1 %
    PVDF, Hastelloy C4, FKM
    max.4 bar
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 470,00 €
    1 470,00 €
    1 * 1 ST
       1 470,00 €